Assay Details
Gene Symbol: LRRC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015287200.2 | XP_015142686.1 | 6 | 4550 | 90 | |
XM_015287202.2 | XP_015142688.1 | 9 | 3533 | 90 | ||
XM_015287207.2 | XP_015142693.1 | 7 | 4792 | 90 | ||
XM_025150987.1 | XP_025006755.1 | 12 | 3734 | 90 | ||
XM_025150988.1 | XP_025006756.1 | 11 | 3648 | 90 | ||
XM_025150990.1 | XP_025006758.1 | 11 | 3689 | 90 | ||
XM_025150991.1 | XP_025006759.1 | 12 | 3691 | 90 | ||
XM_025150992.1 | XP_025006760.1 | 13 | 3911 | 90 | ||
XM_025150993.1 | XP_025006761.1 | 11 | 3609 | 90 | ||
XM_025150994.1 | XP_025006762.1 | 12 | 3829 | 90 | ||
XM_025150995.1 | XP_025006763.1 | 10 | 3533 | 90 | ||
XM_025150996.1 | XP_025006764.1 | 9 | 3407 | 90 | ||
XM_025150997.1 | XP_025006765.1 | 11 | 3605 | 90 | ||
XM_025150998.1 | XP_025006766.1 | 10 | 3494 | 90 | ||
XM_025150999.1 | XP_025006767.1 | 13 | 3626 | 90 | ||
XM_025151000.1 | XP_025006768.1 | 11 | 3704 | 90 | ||
XM_025151001.1 | XP_025006769.1 | 10 | 3428 | 90 | ||
XM_025151002.1 | XP_025006770.1 | 10 | 3529 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
