Assay Details
Gene Symbol: EVA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299781.2 | XP_015155267.1 | 3 - 4 | 628 | 119 | |
XM_015299792.2 | XP_015155278.1 | 3 - 4 | 629 | 119 | ||
XM_015299795.2 | XP_015155281.1 | 3 - 4 | 630 | 119 | ||
XM_015299796.2 | XP_015155282.1 | 3 - 4 | 608 | 119 | ||
XM_025145015.1 | XP_025000783.1 | 3 - 4 | 361 | 119 | ||
XM_416704.6 | XP_416704.1 | 3 - 4 | 607 | 119 | ||
GenBank mRNA | BX931087.1 | - | 3 - 4 | 494 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
