Assay Details
| Gene Symbol: HNRNPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015289581.2 | XP_015145067.1 | 2 - 3 | 265 | 95 | |
| XM_015289582.2 | XP_015145068.1 | 2 - 3 | 249 | 95 | ||
| XM_015289583.2 | XP_015145069.1 | 2 - 3 | 247 | 95 | ||
| XM_015289584.2 | XP_015145070.1 | 2 - 3 | 226 | 95 | ||
| XM_025152381.1 | XP_025008149.1 | 2 - 3 | 217 | 95 | ||
| GenBank mRNA | BX931118.2 | - | 2 - 3 | 227 | 95 | |
| CR353765.1 | - | 2 - 3 | 224 | 95 | ||
More Information
Set Membership: |
|

HNRNPA3
Genomic Map