Assay Details
Gene Symbol: NDUFAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015282937.1 | XP_015138423.1 | 3 - 4 | 412 | 68 | |
XM_025148048.1 | XP_025003816.1 | 3 - 4 | 412 | 68 | ||
XM_025148049.1 | XP_025003817.1 | 3 - 4 | 410 | 68 | ||
GenBank mRNA | BX931199.2 | - | 3 - 4 | 417 | 68 | |
BX934840.1 | - | 3 - 4 | 418 | 68 | ||
BX936105.1 | - | 3 - 4 | 410 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
