Assay Details
| Gene Symbol: RAB34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296088.2 | XP_015151574.1 | 6 - 7 | 680 | 91 | |
| XM_015296089.2 | XP_015151575.1 | 6 - 7 | 680 | 91 | ||
| XM_015296090.2 | XP_015151576.1 | 7 - 8 | 602 | 91 | ||
| XM_015296091.2 | XP_015151577.1 | 7 - 8 | 510 | 91 | ||
| GenBank mRNA | BX931452.2 | - | 7 - 8 | 529 | 91 | |
More Information
Set Membership: |
|

RAB34
Genomic Map