Assay Details
| Gene Symbol: XRCC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280684.2 | XP_015136170.1 | 2 - 3 | 217 | 65 | |
| XM_015280685.2 | XP_015136171.1 | 2 - 3 | 401 | 65 | ||
| XM_025145264.1 | XP_025001032.1 | 2 - 3 | 239 | 65 | ||
| XM_424905.6 | XP_424905.4 | 2 - 3 | 235 | 65 | ||
| GenBank mRNA | BX931641.1 | - | 1 - 2 | 117 | 65 | |
More Information
Set Membership: |
|

XRCC4
Genomic Map