Assay Details
Gene Symbol: LOC101751456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025143530.1 | XP_024999298.1 | 3 - 4 | 385 | 107 |
Gene Symbol: CCT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015298317.2 | XP_015153803.2 | 3 - 4 | 229 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
