Assay Details
| Gene Symbol: ZNF185L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278548.1 | XP_015134034.1 | 33 | 4518 | 104 | |
| XM_015278552.1 | XP_015134038.1 | 32 | 4422 | 104 | ||
| XM_015278553.1 | XP_015134039.1 | 32 | 4416 | 104 | ||
| XM_015278554.1 | XP_015134040.1 | 32 | 4404 | 104 | ||
| XM_015278555.1 | XP_015134041.1 | 31 | 4308 | 104 | ||
| XM_015278556.1 | XP_015134042.1 | 30 | 4206 | 104 | ||
| XM_015278557.1 | XP_015134043.1 | 30 | 4149 | 104 | ||
| XM_025150360.1 | XP_025006128.1 | 32 | 4419 | 104 | ||
| XM_025150361.1 | XP_025006129.1 | 31 | 4302 | 104 | ||
More Information
Set Membership: |
|

ZNF185L
Genomic Map