Assay Details
| Gene Symbol: PLACL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941122.3 | XP_004941179.1 | 1 - 2 | 66 | 84 | |
| Gene Symbol: PLAC8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149758.1 | XP_025005526.1 | 1 - 2 | 81 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004941122.3, XM_025149758.1 transcripts.
Set Membership: |
|

PLACL2
Genomic Map