Assay Details
| Gene Symbol: PLACL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941122.3 | XP_004941179.1 | 4 - 5 | 422 | 97 | |
| Gene Symbol: PLAC8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025149758.1 | XP_025005526.1 | 3 - 4 | 312 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025149758.1 transcripts.
Set Membership: |
|

PLACL2
Genomic Map