Assay Details
| Gene Symbol: RBM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015272997.1 | XP_015128483.1 | 3 | 2618 | 72 | |
| XM_015272998.1 | XP_015128484.1 | 2 | 1006 | 72 | ||
More Information
Set Membership: |
|

RBM14
Genomic Map