Assay Details
| Gene Symbol: PPP1R1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299543.2 | XP_015155029.1 | 1 - 2 | 235 | 109 | |
| XM_424268.6 | XP_424268.3 | 1 - 2 | 181 | 109 | ||
More Information
Set Membership: |
|

PPP1R1B
Genomic Map