Assay Details
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004947168.3 | XP_004947225.1 | 12 - 13 | 1378 | 144 | |
XM_004947170.3 | XP_004947227.1 | 11 - 12 | 1288 | 144 | ||
XM_015296447.2 | XP_015151933.1 | 11 - 12 | 1868 | 144 | ||
XM_015296448.1 | XP_015151934.1 | 12 - 13 | 1407 | 144 | ||
XM_015296449.1 | XP_015151935.1 | 12 - 13 | 1356 | 144 | ||
XM_015296450.2 | XP_015151936.1 | 12 - 13 | 1393 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
