Assay Details
| Gene Symbol: MOBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281725.2 | XP_015137211.1 | 16 - 17 | 2715 | 105 | |
| XM_015281726.2 | XP_015137212.1 | 15 - 16 | 2650 | 105 | ||
| XM_015281727.2 | XP_015137213.1 | 15 - 16 | 2610 | 105 | ||
| XM_025147516.1 | XP_025003284.1 | 14 - 15 | 2321 | 105 | ||
More Information
Set Membership: |
|

MOBP
Genomic Map