Assay Details
| Gene Symbol: NSMCE4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421797.6 | XP_421797.2 | 2 - 3 | 427 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NSMCE4A| Gene Symbol: NSMCE4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421797.6 | XP_421797.2 | 2 - 3 | 427 | 65 | |
Set Membership: |
|