Assay Details
| Gene Symbol: CZH18ORF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004949035.3 | XP_004949092.2 | 4 - 5 | 1210 | 60 | |
| XM_004949037.3 | XP_004949094.2 | 4 - 5 | 1069 | 60 | ||
| XM_004949040.3 | XP_004949097.1 | 3 - 4 | 1018 | 60 | ||
| XM_015277419.2 | XP_015132905.1 | 5 - 6 | 1285 | 60 | ||
| GenBank mRNA | BX931744.2 | - | 2 - 3 | 480 | 60 | |
More Information
Set Membership: |
|

CZH18ORF25
Genomic Map