Assay Details
| Gene Symbol: NDUFA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300117.2 | XP_015155603.1 | 3 - 4 | 276 | 84 | |
| GenBank mRNA | BX931756.2 | - | 3 - 4 | 272 | 84 | |
| BX932151.1 | - | 3 - 4 | 245 | 84 | ||
More Information
Set Membership: |
|

NDUFA13
Genomic Map