Assay Details
| Gene Symbol: RNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282602.2 | XP_015138088.1 | 5 - 6 | 8696 | 72 | |
| XM_015282603.2 | XP_015138089.1 | 5 - 6 | 8779 | 72 | ||
| XM_015282604.2 | XP_015138090.1 | 4 - 5 | 8652 | 72 | ||
| XM_015282605.2 | XP_015138091.1 | 5 - 6 | 8779 | 72 | ||
| XM_015282606.2 | XP_015138092.1 | 4 - 5 | 8652 | 72 | ||
| XM_025147914.1 | XP_025003682.1 | 5 - 6 | 12946 | 72 | ||
| GenBank mRNA | BX932422.2 | - | 3 - 4 | 329 | 72 | |
More Information
Set Membership: |
|

RNF138
Genomic Map