Assay Details
| Gene Symbol: SYNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297850.2 | XP_015153336.1 | 3 - 4 | 1783 | 94 | |
| XM_015297851.2 | XP_015153337.1 | 3 - 4 | 1669 | 94 | ||
| XM_417811.5 | XP_417811.3 | 3 - 4 | 1740 | 94 | ||
More Information
Set Membership: |
|

SYNC
Genomic Map