Assay Details
Gene Symbol: LOC107050760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145636.1 | XP_025001404.1 | 7 - 8 | 784 | 107 | |
GenBank mRNA | KY501151.1 | - | 6 - 7 | 648 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
