Assay Details
| Gene Symbol: MRPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025146646.1 | XP_025002414.1 | 1 - 2 | 67 | 49 | |
| GenBank mRNA | BX932573.2 | - | 1 - 2 | 48 | 49 | |
More Information
Set Membership: |
|

MRPL30
Genomic Map