Assay Details
Gene Symbol: TXNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025143563.1 | XP_024999331.1 | 2 - 3 | 401 | 71 | |
GenBank mRNA | BX933049.1 | - | 2 - 3 | 220 | 71 | |
BX933080.1 | - | 2 - 3 | 129 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
