Assay Details
| Gene Symbol: CDC42EP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015283753.2 | XP_015139239.1 | 1 - 2 | 170 | 130 | |
| GenBank mRNA | BX933559.1 | - | 1 - 2 | 122 | 130 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015283753.2 transcripts.
Set Membership: |
|

CDC42EP3
Genomic Map