Assay Details
| Gene Symbol: SLC30A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642566.4 | XP_003642614.2 | 2 - 3 | 545 | 79 | |
| GenBank mRNA | BX933724.2 | - | 1 - 2 | 208 | 79 | |
More Information
Set Membership: |
|

SLC30A2
Genomic Map