Assay Details
| Gene Symbol: BANF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015274250.2 | XP_015129736.1 | 3 | 1410 | 114 | |
| GenBank mRNA | BX933831.1 | - | 1 | 925 | 114 | |
More Information
Set Membership: |
|

BANF1
Genomic Map