Assay Details
Gene Symbol: PANX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015273068.2 | XP_015128554.1 | 1 - 2 | 394 | 106 | |
XM_015273291.2 | XP_015128777.1 | 1 - 2 | 395 | 106 | ||
GenBank mRNA | BX934189.1 | - | 1 - 2 | 89 | 106 | |
CR405835.1 | - | 1 - 2 | 206 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015273068.2, XM_015273291.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
