Assay Details
Gene Symbol: SEC22C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004939070.3 | XP_004939127.1 | 3 - 4 | 413 | 70 | |
XM_004939071.3 | XP_004939128.1 | 3 - 4 | 838 | 70 | ||
XM_015281147.2 | XP_015136633.1 | 3 - 4 | 1011 | 70 | ||
XM_015281148.2 | XP_015136634.1 | 3 - 4 | 2263 | 70 | ||
XM_025148244.1 | XP_025004012.1 | 3 - 4 | 504 | 70 | ||
XM_418493.6 | XP_418493.1 | 3 - 4 | 2198 | 70 | ||
GenBank mRNA | BX935473.2 | - | 3 - 4 | 397 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
