Assay Details
| Gene Symbol: TPGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642859.4 | XP_003642907.1 | 1 - 2 | 338 | 115 | |
| GenBank mRNA | BX950510.2 | - | 1 - 2 | 329 | 115 | |
More Information
Set Membership: |
|

TPGS1
Genomic Map