Assay Details
| Gene Symbol: RAB18L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_423284.6 | XP_423284.2 | 6 - 7 | 509 | 76 | |
| GenBank mRNA | BX950642.2 | - | 6 - 7 | 493 | 76 | |
More Information
Set Membership: |
|

RAB18L
Genomic Map