Assay Details
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935576.2 | XP_004935633.1 | 2 - 3 | 725 | 95 | |
XM_015284314.2 | XP_015139800.1 | 8 - 9 | 923 | 95 | ||
XM_015284315.2 | XP_015139801.1 | 9 - 10 | 1000 | 95 | ||
XM_015284316.2 | XP_015139802.1 | 7 - 8 | 867 | 95 | ||
XM_025148833.1 | XP_025004601.1 | 8 - 9 | 682 | 95 | ||
GenBank mRNA | BX950648.1 | - | 1 - 2 | 653 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
