Assay Details
| Gene Symbol: AMY1AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003250.2 | 1 - 2 | 865 | 110 | ||
| GenBank mRNA | CR390701.1 | - | 1 - 2 | 865 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AMY1AP| Gene Symbol: AMY1AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003250.2 | 1 - 2 | 865 | 110 | ||
| GenBank mRNA | CR390701.1 | - | 1 - 2 | 865 | 110 | |
Set Membership: |
|