Assay Details
Gene Symbol: AUTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015295943.2 | XP_015151429.1 | 19 | 3791 | 76 | |
XM_015295944.2 | XP_015151430.1 | 19 | 3789 | 76 | ||
XM_015295945.2 | XP_015151431.1 | 18 | 3746 | 76 | ||
XM_015295946.2 | XP_015151432.1 | 18 | 3725 | 76 | ||
XM_015295949.2 | XP_015151435.1 | 14 | 2253 | 76 | ||
XM_415729.6 | XP_415729.2 | 19 | 3770 | 76 | ||
GenBank mRNA | CR389155.1 | - | 2 | 418 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
