Assay Details
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004939985.3 | XP_004940042.2 | 3 | 555 | 75 | |
XM_418371.6 | XP_418371.4 | 3 | 720 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
