Assay Details
Gene Symbol: CZH9orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004937102.3 | XP_004937159.1 | 2 - 3 | 600 | 94 | |
XM_424976.6 | XP_424976.2 | 2 - 3 | 607 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
