Assay Details
| Gene Symbol: PIPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235035.5 | XP_001235036.3 | 6 - 7 | 1010 | 89 | |
| XM_025142086.1 | XP_024997854.1 | 5 - 6 | 862 | 89 | ||
More Information
Set Membership: |
|

PIPOX
Genomic Map