Assay Details
| Gene Symbol: LOC101748581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004934965.3 | XP_004935022.1 | 158 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004934965.3 transcripts.
Set Membership: |
|

LOC101748581
Genomic Map