Assay Details
| Gene Symbol: LOC101748581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004934965.3 | XP_004935022.1 | 2079 | 76 | ||
| XM_025143770.1 | XP_024999538.1 | 2194 | 76 | |||
| XM_025143772.1 | XP_024999540.1 | 2110 | 76 | |||
More Information
Set Membership: |
|

LOC101748581
Genomic Map