Assay Details
Gene Symbol: LRRC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015287708.2 | XP_015143194.1 | 9 - 10 | 1230 | 62 | |
XM_426448.6 | XP_426448.4 | 9 - 10 | 1233 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
