Assay Details
Gene Symbol: LOC101748751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015298099.2 | XP_015153585.1 | 1 - 2 | 236 | 112 | |
XM_015298100.2 | XP_015153586.1 | 1 - 2 | 229 | 112 | ||
XM_015298101.2 | XP_015153587.1 | 1 - 2 | 220 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015298099.2, XM_015298100.2, XM_015298101.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
