Assay Details
Gene Symbol: LOC101748751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015298099.2 | XP_015153585.1 | 3 - 4 | 867 | 65 | |
XM_015298100.2 | XP_015153586.1 | 3 - 4 | 860 | 65 | ||
XM_015298101.2 | XP_015153587.1 | 3 - 4 | 851 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
