Assay Details
Gene Symbol: LOC101748751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015298099.2 | XP_015153585.1 | 6 - 7 | 1699 | 71 | |
XM_015298100.2 | XP_015153586.1 | 5 - 6 | 1398 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
