Assay Details
Gene Symbol: LOC101748751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015298099.2 | XP_015153585.1 | 9 - 10 | 2422 | 55 | |
XM_015298100.2 | XP_015153586.1 | 8 - 9 | 2121 | 55 | ||
XM_015298101.2 | XP_015153587.1 | 7 - 8 | 1851 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
