Assay Details
Gene Symbol: LOC424748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004936949.3 | XP_004937006.1 | 5 - 6 | 731 | 63 | |
XM_025153532.1 | XP_025009300.1 | 5 - 6 | 1936 | 63 | ||
XM_025153533.1 | XP_025009301.1 | 5 - 6 | 544 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
