Assay Details
| Gene Symbol: TRAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296086.2 | XP_015151572.1 | 7 - 8 | 1136 | 92 | |
| XM_025142085.1 | XP_024997853.1 | 7 - 8 | 1243 | 92 | ||
More Information
Set Membership: |
|

TRAF4
Genomic Map