Assay Details
| Gene Symbol: LOC101749673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145791.1 | XP_025001559.1 | 3 - 4 | 1195 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101749673| Gene Symbol: LOC101749673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145791.1 | XP_025001559.1 | 3 - 4 | 1195 | 68 | |
Set Membership: |
|