Assay Details
Gene Symbol: FAM194A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291842.2 | XP_015147328.1 | 10 - 11 | 4047 | 92 | |
XM_015291843.2 | XP_015147329.1 | 11 - 12 | 3957 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
