Assay Details
Gene Symbol: FAM194A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291842.2 | XP_015147328.1 | 1 - 2 | 2938 | 80 | |
XM_015291843.2 | XP_015147329.1 | 2 - 3 | 2848 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
