Assay Details
Gene Symbol: FAM194A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291842.2 | XP_015147328.1 | 6 - 7 | 3422 | 65 | |
XM_015291843.2 | XP_015147329.1 | 7 - 8 | 3332 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
