Assay Details
| Gene Symbol: LOC427775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425346.4 | XP_425346.2 | 2 - 3 | 490 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC427775| Gene Symbol: LOC427775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425346.4 | XP_425346.2 | 2 - 3 | 490 | 57 | |
Set Membership: |
|