Assay Details
| Gene Symbol: SLC2A11L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232863.5 | XP_001232864.2 | 6 - 7 | 753 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC2A11L4| Gene Symbol: SLC2A11L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232863.5 | XP_001232864.2 | 6 - 7 | 753 | 110 | |
Set Membership: |
|